Assay Details
Gene Symbol: DUSP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005673571.2 | XP_005673628.1 | 221 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
