Assay Details
Gene Symbol: BHLHA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013981203.2 | XP_013836657.2 | 1019 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
