Assay Details
| Gene Symbol: FXYD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013983259.2 | XP_013838713.1 | 483 | 67 | ||
| XM_013983261.2 | XP_013838715.1 | 464 | 67 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: FXYD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013983259.2 | XP_013838713.1 | 483 | 67 | ||
| XM_013983261.2 | XP_013838715.1 | 464 | 67 | |||
Set Membership: |
|