Assay Details
Gene Symbol: FZD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013984388.2 | XP_013839842.2 | 1622 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
