Assay Details
Gene Symbol: ECMX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013986548.2 | XP_013842002.1 | 1131 | 79 | ||
XM_021080352.1 | XP_020936011.1 | 2922 | 79 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ECMX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013986548.2 | XP_013842002.1 | 1131 | 79 | ||
XM_021080352.1 | XP_020936011.1 | 2922 | 79 |