Assay Details
Gene Symbol: NRARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013989287.2 | XP_013844741.1 | 616 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
