Assay Details
Gene Symbol: BARHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001929435.5 | XP_001929470.1 | 943 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
