Assay Details
| Gene Symbol: RORB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021064932.1 | XP_020920591.1 | 2663 | 66 | ||
| XM_021064942.1 | XP_020920601.1 | 2657 | 66 | |||
| XM_021064949.1 | XP_020920608.1 | 2455 | 66 | |||
More Information
Set Membership: |
|

RORB
Genomic Map