Assay Details
| Gene Symbol: TAFA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021070829.1 | XP_020926488.1 | 1211 | 64 | ||
| XM_021070830.1 | XP_020926489.1 | 1406 | 64 | |||
| XM_021070831.1 | XP_020926490.1 | 1215 | 64 | |||
More Information
Set Membership: |
|

TAFA1
Genomic Map