Assay Details
Gene Symbol: CALHM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021073163.1 | XP_020928822.1 | 722 | 85 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CALHM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021073163.1 | XP_020928822.1 | 722 | 85 |