Assay Details
| Gene Symbol: LOC100156277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021073254.1 | XP_020928913.1 | 1304 | 76 | ||
| XM_021073255.1 | XP_020928914.1 | 3379 | 76 | |||
More Information
Set Membership: |
|

LOC100156277
Genomic Map