Assay Details
Gene Symbol: TAS2R3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021078592.1 | XP_020934251.1 | 2017 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
