Assay Details
Gene Symbol: LOC102160746 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021082418.1 | XP_020938077.1 | 157 | 52 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021082418.1 transcripts.
Set Membership: |
![]() ![]() |
