Assay Details
Gene Symbol: LOC102160746 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021082418.1 | XP_020938077.1 | 532 | 57 | ||
XM_021082419.1 | XP_020938078.1 | 1814 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
