Assay Details
Gene Symbol: TCP10L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021086007.1 | XP_020941666.1 | 3325 | 86 | ||
XM_021086017.1 | XP_020941676.1 | 638 | 86 | |||
XM_021086023.1 | XP_020941682.1 | 3301 | 86 | |||
XM_021086032.1 | XP_020941691.1 | 3325 | 86 | |||
XM_021086039.1 | XP_020941698.1 | 3325 | 86 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
