Assay Details
Gene Symbol: LOC100524794 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021086494.1 | XP_020942153.1 | 1423 | 128 | ||
XM_021086495.1 | XP_020942154.1 | 1407 | 128 | |||
XM_021086496.1 | XP_020942155.1 | 1420 | 128 | |||
XM_021086497.1 | XP_020942156.1 | 1423 | 128 | |||
XM_021086498.1 | XP_020942157.1 | 1894 | 128 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
