Assay Details
Gene Symbol: PFN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021088369.1 | XP_020944028.1 | 1064 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
