Assay Details
Gene Symbol: NLRP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021095065.1 | XP_020950724.1 | 3130 | 74 | ||
XM_021095066.1 | XP_020950725.1 | 3185 | 74 | |||
XM_021095067.1 | XP_020950726.1 | 3152 | 74 | |||
XM_021095069.1 | XP_020950728.1 | 2959 | 74 |
