Assay Details
Gene Symbol: LOC100525229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021096172.1 | XP_020951831.1 | 2269 | 102 | ||
XM_021096175.1 | XP_020951834.1 | 691 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021096172.1, XM_021096175.1 transcripts.
Set Membership: |
![]() ![]() |
