Assay Details
Gene Symbol: LOC100525229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021096172.1 | XP_020951831.1 | 2598 | 92 | ||
XM_021096173.1 | XP_020951832.1 | 2197 | 92 | |||
XM_021096175.1 | XP_020951834.1 | 1020 | 92 | |||
XM_021096176.1 | XP_020951835.1 | 1106 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
