Assay Details
Gene Symbol: LOC100525229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021096172.1 | XP_020951831.1 | 3002 | 93 | ||
XM_021096173.1 | XP_020951832.1 | 2601 | 93 | |||
XM_021096175.1 | XP_020951834.1 | 1424 | 93 | |||
XM_021096176.1 | XP_020951835.1 | 1510 | 93 |
