Assay Details
Gene Symbol: C6H19orf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021097417.1 | XP_020953076.1 | 2839 | 76 | ||
XM_021097418.1 | XP_020953077.1 | 1070 | 76 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C6H19orf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021097417.1 | XP_020953076.1 | 2839 | 76 | ||
XM_021097418.1 | XP_020953077.1 | 1070 | 76 |