Assay Details
Gene Symbol: CFAP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021098946.1 | XP_020954605.1 | 2120 | 52 | ||
XM_021098947.1 | XP_020954606.1 | 2065 | 52 | |||
XM_021098948.1 | XP_020954607.1 | 2051 | 52 | |||
XM_021098949.1 | XP_020954608.1 | 2048 | 52 | |||
XM_021098950.1 | XP_020954609.1 | 2045 | 52 | |||
XM_021098952.1 | XP_020954611.1 | 1896 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
