Assay Details
Gene Symbol: ADTRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021100189.1 | XP_020955848.1 | 683 | 102 | ||
XM_021100190.1 | XP_020955849.1 | 656 | 102 | |||
XM_021100191.1 | XP_020955850.1 | 686 | 102 | |||
XM_021100192.1 | XP_020955851.1 | 528 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
