Assay Details
| Gene Symbol: PRSS48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021100635.1 | XP_020956294.1 | 50 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021100635.1 transcripts.
Set Membership: |
|

PRSS48
Genomic Map