Assay Details
| Gene Symbol: LOC100513484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021101398.1 | XP_020957057.1 | 387 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021101398.1 transcripts.
Set Membership: |
|

LOC100513484
Genomic Map