Assay Details
Gene Symbol: TAS2R7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003126500.1 | XP_003126548.1 | 419 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
