Assay Details
Gene Symbol: TMEM91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003127157.4 | XP_003127205.1 | 440 | 106 | ||
XM_003355932.4 | XP_003355980.2 | 441 | 106 | |||
XM_021094418.1 | XP_020950077.1 | 440 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
