Assay Details
Gene Symbol: IGSF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003127218.3 | XP_003127266.2 | 911 | 75 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IGSF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003127218.3 | XP_003127266.2 | 911 | 75 |