Assay Details
| Gene Symbol: KIF2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128056.6 | XP_003128104.3 | 1392 | 67 | ||
| XM_005656179.3 | XP_005656236.1 | 1220 | 67 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: KIF2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128056.6 | XP_003128104.3 | 1392 | 67 | ||
| XM_005656179.3 | XP_005656236.1 | 1220 | 67 | |||
Set Membership: |
|