Assay Details
Gene Symbol: MS4A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003353837.5 | XP_003353885.2 | 426 | 128 | ||
XM_013994267.2 | XP_013849721.2 | 551 | 128 | |||
XM_013994268.2 | XP_013849722.2 | 559 | 128 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
