Assay Details
| Gene Symbol: LOC100515185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122044.5 | XP_003122092.2 | 1118 | 88 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100515185| Gene Symbol: LOC100515185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122044.5 | XP_003122092.2 | 1118 | 88 | ||
Set Membership: |
|