Assay Details
Gene Symbol: CDRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013980267.2 | XP_013835721.1 | 1649 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013980267.2 transcripts.
Set Membership: |
![]() ![]() |
