Assay Details
| Gene Symbol: NSMCE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021088934.1 | XP_020944593.1 | 864 | 65 | ||
| XM_021088935.1 | XP_020944594.1 | 853 | 65 | |||
| XM_021088936.1 | XP_020944595.1 | 861 | 65 | |||
| XM_021088937.1 | XP_020944596.1 | 779 | 65 | |||
| XM_021088938.1 | XP_020944597.1 | 769 | 65 | |||
| XM_021088939.1 | XP_020944598.1 | 773 | 65 | |||
| XM_021088941.1 | XP_020944600.1 | 659 | 65 | |||
More Information
Set Membership: |
|

NSMCE2
Genomic Map