Assay Details
| Gene Symbol: NSMCE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021088937.1 | XP_020944596.1 | 578 | 74 | ||
| XM_021088938.1 | XP_020944597.1 | 568 | 74 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: NSMCE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021088937.1 | XP_020944596.1 | 578 | 74 | ||
| XM_021088938.1 | XP_020944597.1 | 568 | 74 | |||