Assay Details
Gene Symbol: CA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001927805.2 | XP_001927840.1 | 913 | 68 | ||
GenBank mRNA | AK232899.1 | - | 6 - 7 | 735 | 68 | |
AK392291.1 | - | 6 - 7 | 791 | 68 | ||
AK399951.1 | - | 6 - 7 | 890 | 68 | ||
AK400468.1 | - | 6 - 7 | 735 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
