Assay Details
Gene Symbol: TARBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003126194.6 | XP_003126242.1 | 1246 | 57 | ||
XM_003355410.4 | XP_003355458.1 | 1050 | 57 | |||
XM_021091716.1 | XP_020947375.1 | 967 | 57 | |||
XM_021091718.1 | XP_020947377.1 | 847 | 57 | |||
GenBank mRNA | AK232920.1 | - | 8 - 9 | 1004 | 57 | |
AK389528.1 | - | 8 - 9 | 1044 | 57 | ||
AK398430.1 | - | 4 - 5 | 895 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
