Assay Details
| Gene Symbol: PPP1R3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK232938.1 | - | 1 | 1920 | 92 | |
| AK398949.1 | - | 1 | 1899 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP1R3B| Gene Symbol: PPP1R3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK232938.1 | - | 1 | 1920 | 92 | |
| AK398949.1 | - | 1 | 1899 | 92 | ||
Set Membership: |
|