Assay Details
Gene Symbol: RETREG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001925566.5 | XP_001925601.3 | 1156 | 91 | ||
XM_021074926.1 | XP_020930585.1 | 1059 | 91 | |||
XM_021074927.1 | XP_020930586.1 | 833 | 91 | |||
GenBank mRNA | AK233040.1 | - | 1 - 2 | 119 | 91 | |
AK234653.1 | - | 2 - 3 | 241 | 91 | ||
AK346271.1 | - | 8 - 9 | 1088 | 91 | ||
AK393552.1 | - | 8 - 9 | 1077 | 91 | ||
AK398254.1 | - | 3 - 4 | 285 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
