Assay Details
| Gene Symbol: TXNDC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096661.1 | XP_020952320.1 | 613 | 77 | ||
| GenBank mRNA | AK233063.1 | - | 1 - 2 | 199 | 77 | |
| AK235596.1 | - | 1 - 2 | 199 | 77 | ||
| AK389217.1 | - | 1 - 2 | 199 | 77 | ||
| AK397521.1 | - | 1 - 2 | 199 | 77 | ||
More Information
Set Membership: |
|

TXNDC12
Genomic Map