Assay Details
| Gene Symbol: NUDT17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK233153.1 | - | 1 | 317 | 76 | |
| AK396976.1 | - | 1 | 682 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: NUDT17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK233153.1 | - | 1 | 317 | 76 | |
| AK396976.1 | - | 1 | 682 | 76 | ||
Set Membership: |
|