Assay Details
| Gene Symbol: OCEL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123490.4 | XP_003123538.1 | 660 | 53 | ||
| XM_005661185.3 | XP_005661242.1 | 970 | 53 | |||
| XM_005661187.3 | XP_005661244.1 | 969 | 53 | |||
| XM_005661188.3 | XP_005661245.1 | 660 | 53 | |||
| GenBank mRNA | AK233228.1 | - | 2 - 3 | 1097 | 53 | |
| AK234330.1 | - | 4 - 5 | 644 | 53 | ||
| AK347622.1 | - | 3 - 4 | 770 | 53 | ||
| AK350595.1 | - | 3 - 4 | 558 | 53 | ||
| AK350889.1 | - | 3 - 4 | 634 | 53 | ||
| AK394013.1 | - | 4 - 5 | 629 | 53 | ||
| AK397579.1 | - | 2 - 3 | 713 | 53 | ||
More Information
Set Membership: |
|

OCEL1
Genomic Map