Assay Details
| Gene Symbol: FBXW4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924973.5 | XP_001925008.4 | 1079 | 103 | ||
| XM_021072714.1 | XP_020928373.1 | 1095 | 103 | |||
| XM_021072715.1 | XP_020928374.1 | 1102 | 103 | |||
| XM_021072716.1 | XP_020928375.1 | 1080 | 103 | |||
| XM_021072718.1 | XP_020928377.1 | 1961 | 103 | |||
| XM_021072719.1 | XP_020928378.1 | 1960 | 103 | |||
| GenBank mRNA | AK233258.1 | - | 3 - 4 | 420 | 103 | |
| AK347316.1 | - | 1 - 2 | 41 | 103 | ||
| AK389303.1 | - | 3 - 4 | 689 | 103 | ||
More Information
Set Membership: |
|

FBXW4
Genomic Map