Assay Details
Gene Symbol: MED19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003122715.5 | XP_003122763.3 | 651 | 65 | ||
GenBank mRNA | AK233271.1 | - | 2 - 3 | 504 | 65 | |
AK392961.1 | - | 2 - 3 | 506 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
