Assay Details
Gene Symbol: ZNF207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005656982.3 | XP_005657039.1 | 637 | 56 | ||
XM_005656983.3 | XP_005657040.1 | 637 | 56 | |||
XM_005656984.3 | XP_005657041.1 | 634 | 56 | |||
XM_013989956.2 | XP_013845410.1 | 634 | 56 | |||
XM_013989957.2 | XP_013845411.1 | 634 | 56 | |||
XM_021067451.1 | XP_020923110.1 | 638 | 56 | |||
XM_021067452.1 | XP_020923111.1 | 638 | 56 | |||
XM_021067453.1 | XP_020923112.1 | 634 | 56 | |||
GenBank mRNA | AK233284.1 | - | 4 - 5 | 600 | 56 | |
AK389634.1 | - | 4 - 5 | 56 | |||
AK391525.1 | - | 4 - 5 | 56 | |||
AK396030.1 | - | 4 - 5 | 56 | |||
AK396095.1 | - | 4 - 5 | 602 | 56 | ||
AK396310.1 | - | 4 - 5 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
