Assay Details
| Gene Symbol: SZRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005665044.3 | XP_005665101.1 | 428 | 73 | ||
| GenBank mRNA | AK233556.1 | - | 2 - 3 | 322 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SZRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005665044.3 | XP_005665101.1 | 428 | 73 | ||
| GenBank mRNA | AK233556.1 | - | 2 - 3 | 322 | 73 | |
Set Membership: |
|