Assay Details
Gene Symbol: LOC100520216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013980819.2 | XP_013836273.2 | 512 | 89 | ||
XM_021068115.1 | XP_020923774.1 | 625 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013980819.2, XM_021068115.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
