Assay Details
Gene Symbol: LOC100520216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013980819.2 | XP_013836273.2 | 984 | 79 | ||
XM_021068115.1 | XP_020923774.1 | 1097 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
