Assay Details
Gene Symbol: FCER1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021089597.1 | XP_020945256.1 | 416 | 97 | ||
XM_021089598.1 | XP_020945257.1 | 395 | 97 | |||
GenBank mRNA | KU527059.1 | - | 1 - 2 | 273 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
