Assay Details
| Gene Symbol: CHST12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021085992.1 | XP_020941651.1 | 112 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021085992.1 transcripts.
Set Membership: |
|

CHST12
Genomic Map